Assay Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 19 - 20 | 2128 | 95 | |
| GenBank mRNA | AB037111.1 | - | 20 - 21 | 2413 | 95 | |
| AB037181.1 | - | 19 - 20 | 2128 | 95 | ||
| AK080951.1 | - | 19 - 20 | 2305 | 95 | ||
| BC022604.1 | - | 10 - 11 | 910 | 95 | ||
| BC137898.1 | - | 19 - 20 | 2111 | 8860917 | 95 | |
| BC145016.1 | - | 19 - 20 | 2111 | 9053564 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah2
Genomic Map