Assay Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 22 - 23 | 2437 | 66 | |
| GenBank mRNA | AB037111.1 | - | 23 - 24 | 2722 | 66 | |
| AB037181.1 | - | 22 - 23 | 2437 | 66 | ||
| AK080951.1 | - | 22 - 23 | 2614 | 66 | ||
| BC022604.1 | - | 13 - 14 | 1219 | 66 | ||
| BC137898.1 | - | 22 - 23 | 2420 | 8860917 | 66 | |
| BC145016.1 | - | 22 - 23 | 2420 | 9053564 | 66 | |
More Information
Set Membership: |
|

Asah2
Genomic Map