Assay Details
| Gene Symbol: Akr1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018859.2 | NP_061347.2 | 1 - 2 | 81 | 100 | |
| GenBank mRNA | AI893997.1 | - | 1 - 2 | 81 | 100 | |
| AK002507.1 | - | 1 - 2 | 56 | 100 | ||
| AK007924.1 | - | 1 - 2 | 58 | 100 | ||
| BC012692.1 | - | 1 - 2 | 97 | 3986863 | 100 | |
| U68535.1 | - | 1 - 2 | 67 | 100 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK002507.1,BC012692.1,NM_018859.2,U68535.1 transcripts.
Set Membership: |
|

Akr1e1
Genomic Map