Assay Details
| Gene Symbol: Srcin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018873.2 | NP_061361.2 | 6 - 7 | 984 | 81 | |
| GenBank mRNA | AF040944.1 | - | 6 - 7 | 989 | 81 | |
| AK129422.1 | - | 1 - 2 | 141 | 81 | ||
| CJ061326.1 | - | 2 - 3 | 388 | 81 | ||
| U59873.1 | - | 6 - 7 | 988 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srcin1
Genomic Map