Assay Details
| Gene Symbol: Srcin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018873.2 | NP_061361.2 | 16 - 17 | 3202 | 62 | |
| GenBank mRNA | AF040944.1 | - | 16 - 17 | 3204 | 62 | |
| AK129422.1 | - | 11 - 12 | 2359 | 62 | ||
| U59873.1 | - | 16 - 17 | 3203 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srcin1
Genomic Map