Assay Details
| Gene Symbol: Srcin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018873.2 | NP_061361.2 | 18 - 19 | 3507 | 55 | |
| GenBank mRNA | AF040944.1 | - | 18 - 19 | 3509 | 55 | |
| AK129422.1 | - | 13 - 14 | 2664 | 55 | ||
| U59873.1 | - | 18 - 19 | 3508 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srcin1
Genomic Map