Assay Details
| Gene Symbol: Paxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018878.3 | NP_061366.2 | 15 - 16 | 2879 | 68 | |
| GenBank mRNA | AF104261.1 | - | 15 - 16 | 2879 | 68 | |
| AK144606.1 | - | 15 - 16 | 2741 | 68 | ||
| AK148141.1 | - | 15 - 16 | 2751 | 68 | ||
| BC066014.1 | - | 15 - 16 | 2668 | 5704566 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paxip1
Genomic Map