Assay Details
| Gene Symbol: Nfkb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177369.1 | NP_001170840.1 | 5 - 6 | 394 | 60 | |
| NM_001177370.1 | NP_001170841.1 | 4 - 5 | 310 | 60 | ||
| NM_019408.3 | NP_062281.1 | 4 - 5 | 545 | 60 | ||
| GenBank mRNA | AF155372.1 | - | 5 - 6 | 472 | 60 | |
| AF155373.1 | - | 5 - 6 | 509 | 60 | ||
| AK134081.1 | - | 5 - 6 | 813 | 60 | ||
| AK137680.1 | - | 5 - 6 | 384 | 60 | ||
| AK144994.1 | - | 5 - 6 | 387 | 60 | ||
| AK145312.1 | - | 4 - 5 | 511 | 60 | ||
| AK148160.1 | - | 5 - 6 | 382 | 60 | ||
| AK150012.1 | - | 5 - 6 | 373 | 60 | ||
| BC027423.1 | - | 4 - 5 | 311 | 3672913 | 60 | |
| CJ134893.1 | - | 4 - 5 | 542 | 60 | ||
| DQ020182.1 | - | 4 - 5 | 242 | 60 | ||
| DQ020183.1 | - | 3 - 4 | 363 | 60 | ||
| DQ020184.1 | - | 3 - 4 | 363 | 60 | ||
| DQ020185.1 | - | 4 - 5 | 242 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkb2
Genomic Map