Assay Details
| Gene Symbol: Ppp2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019411.4 | NP_062284.1 | 2 - 3 | 537 | 104 | |
| GenBank mRNA | AF076192.1 | - | 2 - 3 | 507 | 104 | |
| AK076110.1 | - | 2 - 3 | 538 | 104 | ||
| AK172644.1 | - | 2 - 3 | 510 | 104 | ||
| AK199825.1 | - | 1 - 2 | 181 | 104 | ||
| BC003856.1 | - | 2 - 3 | 514 | 3484084 | 104 | |
| BC054458.1 | - | 2 - 3 | 497 | 6493866 | 104 | |
| Z67745.1 | - | 2 - 3 | 104 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2ca
Genomic Map