Assay Details
| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019426.2 | NP_062299.2 | 1 - 2 | 110 | 109 | |
| GenBank mRNA | AJ132702.1 | - | 1 - 2 | 103 | 109 | |
| AK082697.1 | - | 1 - 2 | 160 | 109 | ||
| AK134252.1 | - | 1 - 2 | 117 | 109 | ||
| AK167542.1 | - | 1 - 2 | 109 | |||
| BC022714.1 | - | 1 - 2 | 103 | 109 | ||
| BC052460.1 | - | 1 - 2 | 110 | 6834550 | 109 | |
| BC060246.1 | - | 1 - 2 | 93 | 5705945 | 109 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ132702.1,AK082697.1,AK134252.1,AK167542.1,BC022714.1,BC052460.1,BC060246.1,NM_019426.2 transcripts.
Set Membership: |
|

Atf7ip
Genomic Map