Assay Details
| Gene Symbol: Nmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019515.1 | NP_062388.1 | 1 - 2 | 220 | 101 | |
| GenBank mRNA | AF203444.1 | - | 1 - 2 | 220 | 101 | |
| AK077478.1 | - | 1 - 2 | 232 | 101 | ||
| BC125545.1 | - | 1 - 2 | 185 | 40130060 | 101 | |
| BC138071.1 | - | 1 - 2 | 185 | 8861090 | 101 | |
| BC145135.1 | - | 1 - 2 | 185 | 9053683 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmu
Genomic Map