Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600007213.mm.1 |
TC0600000941.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007213.mm.1 |
NM_001164187.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007213.mm.1 |
NM_001164187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007213.mm.1 |
NM_019542.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007213.mm.1 |
NM_019542.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007209.mm.1 |
TC0600000941.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007209.mm.1 |
NM_019542.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007209.mm.1 |
NM_019542.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007211.mm.1 |
TC0600000941.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007211.mm.1 |
NM_001164187.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007211.mm.1 |
NM_001164187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007211.mm.1 |
NM_019542.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007211.mm.1 |
NM_019542.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007215.mm.1 |
TC0600000941.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007215.mm.1 |
NM_001164187.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007215.mm.1 |
NM_001164187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007215.mm.1 |
NM_019542.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007215.mm.1 |
NM_019542.2 |
Refseq |