Assay Details
| Gene Symbol: Htra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019564.3 | NP_062510.2 | 4 - 5 | 1040 | 73 | |
| GenBank mRNA | AF172994.1 | - | 4 - 5 | 1016 | 73 | |
| AF179369.1 | - | 4 - 5 | 1011 | 73 | ||
| AK090320.1 | - | 4 - 5 | 755 | 73 | ||
| AK090321.1 | - | 1 - 2 | 187 | 73 | ||
| AK128916.1 | - | 4 - 5 | 1013 | 73 | ||
| AK132270.1 | - | 4 - 5 | 1013 | 73 | ||
| AK167667.1 | - | 4 - 5 | 606 | 73 | ||
| BC013516.1 | - | 4 - 5 | 1010 | 4235912 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Htra1
Genomic Map