Assay Details
| Gene Symbol: Hsd17b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019657.4 | NP_062631.1 | 9 - 10 | 720 | 117 | |
| GenBank mRNA | AF064635.1 | - | 9 - 10 | 699 | 117 | |
| AK012103.1 | - | 2 - 3 | 134 | 117 | ||
| AK078841.1 | - | 9 - 10 | 721 | 117 | ||
| AK081869.1 | - | 9 - 10 | 714 | 117 | ||
| AK082715.1 | - | 9 - 10 | 714 | 117 | ||
| AK088521.1 | - | 9 - 10 | 714 | 117 | ||
| AK150849.1 | - | 9 - 10 | 715 | 117 | ||
| AK152489.1 | - | 9 - 10 | 715 | 117 | ||
| AK166027.1 | - | 9 - 10 | 714 | 117 | ||
| AK167908.1 | - | 9 - 10 | 714 | 117 | ||
| AK188070.1 | - | 4 - 5 | 197 | 117 | ||
| BC010749.1 | - | 9 - 10 | 695 | 117 | ||
| BC037620.1 | - | 8 - 9 | 637 | 5375831 | 117 | |
| BC090659.1 | - | 9 - 10 | 690 | 30675153 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b12
Genomic Map