Assay Details
| Gene Symbol: Actl6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019673.2 | NP_062647.2 | 1 - 2 | 328 | 87 | |
| GenBank mRNA | AF041476.1 | - | 1 - 2 | 148 | 87 | |
| AK002724.1 | - | 1 - 2 | 124 | 87 | ||
| AK090154.1 | - | 1 - 2 | 147 | 87 | ||
| AK167905.1 | - | 1 - 2 | 142 | 87 | ||
| AK169278.1 | - | 1 - 2 | 147 | 87 | ||
| BC001994.1 | - | 1 - 2 | 116 | 3491205 | 87 | |
| BC094498.1 | - | 1 - 2 | 144 | 4209491 | 87 | |
| CF725603.1 | - | 1 - 2 | 328 | 87 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF041476.1,AK090154.1,AK167905.1,AK169278.1,BC001994.1,BC094498.1,NM_019673.2 transcripts.
Set Membership: |
|

Actl6a
Genomic Map