Assay Details
| Gene Symbol: Actl6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019673.2 | NP_062647.2 | 2 - 3 | 415 | 72 | |
| GenBank mRNA | AF041476.1 | - | 2 - 3 | 235 | 72 | |
| AK002724.1 | - | 2 - 3 | 211 | 72 | ||
| AK090154.1 | - | 2 - 3 | 72 | |||
| AK167905.1 | - | 2 - 3 | 229 | 72 | ||
| AK169278.1 | - | 2 - 3 | 234 | 72 | ||
| AK182047.1 | - | 1 - 2 | 71 | |||
| AK205745.1 | - | 1 - 2 | 73 | 72 | ||
| BC001994.1 | - | 2 - 3 | 203 | 3491205 | 72 | |
| BC094498.1 | - | 2 - 3 | 231 | 4209491 | 72 | |
| CF725603.1 | - | 2 - 3 | 415 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actl6a
Genomic Map