| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700035942.mm.1 |
NM_019674.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700035942.mm.1 |
NM_019674.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700035942.mm.1 |
TC0700004295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700035941.mm.1 |
NM_019674.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700035941.mm.1 |
NM_019674.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700035941.mm.1 |
TC0700004295.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700035940.mm.1 |
NM_019674.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700035940.mm.1 |
NM_019674.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700035940.mm.1 |
TC0700004295.mm.1 |
MTA TC |