Assay Details
| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019676.3 | NP_062650.1 | 1 - 2 | 145 | 55 | |
| NR_121571.1 | 1 - 2 | 145 | 55 | |||
| GenBank mRNA | AF133125.1 | - | 1 - 2 | 127 | 55 | |
| AK028749.1 | - | 1 - 2 | 146 | 55 | ||
| AK082890.1 | - | 1 - 2 | 137 | 55 | ||
| BC015249.1 | - | 1 - 2 | 99 | 4241580 | 55 | |
| BC025798.1 | - | 1 - 2 | 121 | 5068610 | 55 | |
| U85711.1 | - | 1 - 2 | 139 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plcd1
Genomic Map