Assay Details
| Gene Symbol: Arl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019722.3 | NP_062696.2 | 2 - 3 | 216 | 70 | |
| GenBank mRNA | AF143680.1 | - | 2 - 3 | 180 | 70 | |
| AK011366.1 | - | 2 - 3 | 216 | 70 | ||
| BC052938.1 | - | 2 - 3 | 195 | 70 | ||
| BC060259.1 | - | 2 - 3 | 206 | 5709669 | 70 | |
| BC080803.1 | - | 2 - 3 | 219 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arl2
Genomic Map