Assay Details
| Gene Symbol: Nat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019750.3 | NP_062724.1 | 1 - 2 | 768 | 63 | |
| GenBank mRNA | AF172275.1 | - | 1 - 2 | 148 | 63 | |
| AK137391.1 | - | 1 - 2 | 134 | 63 | ||
| BC026545.1 | - | 1 - 2 | 793 | 5052098 | 63 | |
| Gene Symbol: Hyal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF417494.1 | - | 851 | 63 | ||
| AF417498.1 | - | 1924 | 63 | |||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF172275.1,AK137391.1,BC026545.1,NM_019750.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
biological_process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P268145 | BC026545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P268145 | NM_019750.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P268145 | AF172275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P268145 | AK137391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P268145 | BC026545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P268145 | NM_019750.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P268145 | AF172275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P268145 | AK137391.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965366 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340706 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65907 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P268145 | BC026545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P268145 | NM_019750.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P268145 | AF172275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P268145 | AK137391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P268145 | BC026545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P268145 | NM_019750.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P268145 | AF172275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P268145 | AK137391.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268145 | BC026545.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268145 | NM_019750.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268145 | AF172275.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268145 | AK137391.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268145 | BC026545.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268145 | NM_019750.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268145 | AF172275.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268145 | AK137391.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P179047 | BC026545.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P179047 | NM_019750.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P179047 | AF172275.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26809711 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02024842 | - | - |
| Whole Mouse Genome G4122A | A_52_P179047 | BC026545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P179047 | NM_019750.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P179047 | AF172275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268145 | BC026545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268145 | NM_019750.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268145 | AF172275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268145 | AK137391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268145 | BC026545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268145 | NM_019750.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268145 | AF172275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268145 | AK137391.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07582771 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07582769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809770 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588695 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588694 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588697 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588700 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588698 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588699 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419213_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588696 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588691 | - | - |
| Mouse Genome 430 2.0 Array | 1419213_at | - | - |
| Mouse Genome 430 2.0 Array | 1419213_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56441_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56441_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900010842.mm.1 | NM_019750.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010842.mm.1 | TC0900001376.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010833.mm.1 | NM_019750.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010833.mm.1 | TC0900001376.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010832.mm.1 | NM_019750.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010832.mm.1 | TC0900001376.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010834.mm.1 | NM_019750.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010834.mm.1 | TC0900001376.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010841.mm.1 | NM_019750.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010841.mm.1 | TC0900001376.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449954_at | - | - |
| Mouse Expression Set 430 | 1449954_PM_at | - | - |
| Mouse Expression Set 430 | 1419213_at | - | - |
| Mouse Expression Set 430 | 1419213_PM_at | - | - |
| Murine Genome U74v2 Set | 104439_PM_at | - | - |
| Murine Genome U74v2 Set | 102230_at | - | - |
| Murine Genome U74v2 Set | 104439_at | - | - |
| Murine Genome U74v2 Set | 102230_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419213_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419213_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449954_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449954_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419213_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419213_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 440835 | - | - |
| ABI Mouse Genome Survey Microarray | 452557 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1050039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708415 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2708415 | - | - |
| mouse-6 v1.1 (Illumina) | scl056441.3_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056441.3_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2905859 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2708415 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2905859 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708415 | - | - |

Nat6
Genomic Map