Assay Details
| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177983.1 | NP_001171454.1 | 2 - 3 | 914 | 74 | |
| NM_019840.2 | NP_062814.2 | 2 - 3 | 717 | 74 | ||
| GenBank mRNA | AF208023.2 | - | 2 - 3 | 652 | 74 | |
| AF326555.1 | - | 2 - 3 | 457 | 74 | ||
| AF326556.1 | - | 2 - 3 | 457 | 74 | ||
| AJ297397.1 | - | 2 - 3 | 652 | 74 | ||
| BC049864.1 | - | 2 - 3 | 915 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde4b
Genomic Map