Assay Details
| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177983.1 | NP_001171454.1 | 4 - 5 | 1022 | 67 | |
| NM_019840.2 | NP_062814.2 | 4 - 5 | 825 | 67 | ||
| GenBank mRNA | AF208023.2 | - | 4 - 5 | 760 | 67 | |
| AF326555.1 | - | 4 - 5 | 565 | 67 | ||
| AF326556.1 | - | 4 - 5 | 565 | 67 | ||
| AJ297397.1 | - | 4 - 5 | 760 | 67 | ||
| AK149150.1 | - | 5 - 6 | 670 | 67 | ||
| BC049864.1 | - | 4 - 5 | 1023 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde4b
Genomic Map