Assay Details
| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177980.1 | NP_001171451.1 | 5 - 6 | 1356 | 67 | |
| NM_001177981.1 | NP_001171452.1 | 5 - 6 | 896 | 67 | ||
| NM_001177982.1 | NP_001171453.1 | 5 - 6 | 632 | 67 | ||
| NM_019840.2 | NP_062814.2 | 10 - 11 | 1521 | 67 | ||
| GenBank mRNA | AF208023.2 | - | 10 - 11 | 1456 | 67 | |
| AF326555.1 | - | 10 - 11 | 1261 | 67 | ||
| AF326556.1 | - | 10 - 11 | 1261 | 67 | ||
| AJ297397.1 | - | 10 - 11 | 1456 | 67 | ||
| AK051102.1 | - | 5 - 6 | 897 | 67 | ||
| AK161341.1 | - | 5 - 6 | 633 | 67 | ||
| BC007155.1 | - | 5 - 6 | 703 | 67 | ||
| BC023751.1 | - | 5 - 6 | 1332 | 67 | ||
| BC071259.1 | - | 5 - 6 | 872 | 5368075 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde4b
Genomic Map