Assay Details
| Gene Symbol: Tmem9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020050.1 | NP_064434.1 | 2 - 3 | 288 | 84 | |
| GenBank mRNA | AK007430.1 | - | 2 - 3 | 256 | 84 | |
| AK009147.1 | - | 2 - 3 | 223 | 84 | ||
| AK049882.1 | - | 2 - 3 | 375 | 84 | ||
| AK077253.1 | - | 2 - 3 | 244 | 84 | ||
| AK147127.1 | - | 2 - 3 | 304 | 84 | ||
| AK201035.1 | - | 1 - 2 | 102 | 84 | ||
| BC002208.1 | - | 2 - 3 | 272 | 3490055 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem9b
Genomic Map