Assay Details
| Gene Symbol: Brd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286630.1 | NP_001273559.1 | 3 - 4 | 593 | 84 | |
| NM_020508.4 | NP_065254.3 | 3 - 4 | 549 | 84 | ||
| NM_198094.2 | NP_932762.2 | 3 - 4 | 539 | 84 | ||
| GenBank mRNA | AF273217.1 | - | 2 - 3 | 464 | 84 | |
| AF461395.1 | - | 3 - 4 | 525 | 84 | ||
| AF461396.1 | - | 3 - 4 | 525 | 84 | ||
| AK042431.1 | - | 3 - 4 | 639 | 84 | ||
| AK147545.1 | - | 3 - 4 | 558 | 84 | ||
| AK154514.1 | - | 2 - 3 | 524 | 84 | ||
| AK184145.1 | - | 1 - 2 | 69 | 84 | ||
| BC011541.1 | - | 2 - 3 | 385 | 84 | ||
| BC013666.1 | - | 2 - 3 | 522 | 84 | ||
| BC082782.1 | - | 2 - 3 | 337 | 84 | ||
| BC103773.1 | - | 2 - 3 | 463 | 84 | ||
| BC138834.1 | - | 3 - 4 | 519 | 8861856 | 84 | |
| BC138835.1 | - | 3 - 4 | 518 | 8861857 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brd4
Genomic Map