Assay Details
| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020606.5 | NP_065631.3 | 11 - 12 | 1232 | 61 | |
| GenBank mRNA | AB045321.1 | - | 11 - 12 | 1105 | 61 | |
| AF237774.1 | - | 11 - 12 | 1105 | 61 | ||
| AF264766.1 | - | 11 - 12 | 1002 | 61 | ||
| AK076473.1 | - | 11 - 12 | 1020 | 61 | ||
| AK077368.1 | - | 12 - 13 | 1064 | 61 | ||
| AK148878.1 | - | 11 - 12 | 966 | 61 | ||
| AK193636.1 | - | 2 - 3 | 165 | 61 | ||
| BC039750.1 | - | 11 - 12 | 990 | 61 | ||
| BC059236.1 | - | 11 - 12 | 1232 | 6400393 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parva
Genomic Map