Assay Details
| Gene Symbol: Ap4m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021392.4 | NP_067367.3 | 2 - 3 | 311 | 111 | |
| GenBank mRNA | AF242858.1 | - | 2 - 3 | 179 | 111 | |
| AK031638.1 | - | 2 - 3 | 314 | 111 | ||
| BC011174.1 | - | 2 - 3 | 196 | 4207033 | 111 | |
| CJ159660.1 | - | 2 - 3 | 331 | 111 | ||
More Information
Set Membership: |
|

Ap4m1
Genomic Map