Assay Details
| Gene Symbol: Ap4m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021392.4 | NP_067367.3 | 9 - 10 | 904 | 137 | |
| GenBank mRNA | AF242858.1 | - | 9 - 10 | 772 | 137 | |
| BC011174.1 | - | 9 - 10 | 789 | 4207033 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap4m1
Genomic Map