Assay Details
| Gene Symbol: Nelfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021393.2 | NP_067368.2 | 2 - 3 | 553 | 93 | |
| GenBank mRNA | AB041607.1 | - | 1 - 2 | 674 | 93 | |
| AK080419.1 | - | 2 - 3 | 255 | 93 | ||
| AK087524.1 | - | 2 - 3 | 253 | 93 | ||
| AK173124.1 | - | 1 - 2 | 88 | 93 | ||
| AK179858.1 | - | 1 - 2 | 60 | 93 | ||
| BC004762.1 | - | 2 - 3 | 441 | 3155904 | 93 | |
| BU519080.1 | - | 2 - 3 | 486 | 93 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB041607.1,BC004762.1 transcripts.
Set Membership: |
|

Nelfb
Genomic Map