Assay Details
| Gene Symbol: Nelfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021393.2 | NP_067368.2 | 6 - 7 | 1179 | 57 | |
| GenBank mRNA | AB041607.1 | - | 5 - 6 | 1300 | 57 | |
| AK080419.1 | - | 6 - 7 | 881 | 57 | ||
| AK087524.1 | - | 6 - 7 | 879 | 57 | ||
| AK173124.1 | - | 5 - 6 | 714 | 57 | ||
| AK179149.1 | - | 1 - 2 | 53 | 57 | ||
| AK189417.1 | - | 1 - 2 | 53 | 57 | ||
| AK214584.1 | - | 1 - 2 | 53 | 57 | ||
| BC004762.1 | - | 6 - 7 | 1067 | 3155904 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nelfb
Genomic Map