Assay Details
| Gene Symbol: Moxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021509.5 | NP_067484.2 | 10 - 11 | 1593 | 73 | |
| GenBank mRNA | AB041606.1 | - | 10 - 11 | 1528 | 73 | |
| AK014340.1 | - | 10 - 11 | 1594 | 73 | ||
| AK030274.1 | - | 10 - 11 | 1534 | 73 | ||
| AK162030.1 | - | 10 - 11 | 1535 | 73 | ||
| BC025892.1 | - | 10 - 11 | 1291 | 73 | ||
| BC037684.1 | - | 10 - 11 | 1531 | 73 | ||
| BC057652.1 | - | 10 - 11 | 1507 | 4017199 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Moxd1
Genomic Map