Assay Details
| Gene Symbol: Slc22a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021551.4 | NP_067526.2 | 7 - 8 | 1549 | 103 | |
| GenBank mRNA | AB012808.1 | - | 9 - 10 | 1335 | 103 | |
| AK007059.1 | - | 7 - 8 | 1494 | 103 | ||
| AK029480.1 | - | 1 - 2 | 158 | 103 | ||
| BC062878.1 | - | 7 - 8 | 1568 | 5696414 | 103 | |
More Information
Set Membership: |
|

Slc22a17
Genomic Map