Assay Details
| Gene Symbol: Nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021605.3 | NP_067618.1 | 7 - 8 | 763 | 126 | |
| GenBank mRNA | AF217650.1 | - | 6 - 7 | 614 | 126 | |
| AK035502.1 | - | 7 - 8 | 762 | 126 | ||
| AK088173.1 | - | 7 - 8 | 812 | 126 | ||
| AK149308.1 | - | 7 - 8 | 811 | 126 | ||
| BC026541.1 | - | 7 - 8 | 755 | 124 | ||
| BC037697.1 | - | 7 - 8 | 740 | 3994871 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek7
Genomic Map