Assay Details
| Gene Symbol: Nek6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159631.1 | NP_001153103.1 | 3 - 4 | 365 | 59 | |
| NM_021606.3 | NP_067619.1 | 3 - 4 | 358 | 59 | ||
| GenBank mRNA | AF218847.1 | - | 3 - 4 | 360 | 59 | |
| AK004925.1 | - | 3 - 4 | 335 | 59 | ||
| AK011519.1 | - | 3 - 4 | 344 | 59 | ||
| AK018168.1 | - | 3 - 4 | 348 | 59 | ||
| AK029266.1 | - | 3 - 4 | 347 | 59 | ||
| AK075717.1 | - | 3 - 4 | 332 | 59 | ||
| AK075836.1 | - | 3 - 4 | 344 | 59 | ||
| AK086700.1 | - | 363 | 59 | |||
| AK089919.1 | - | 3 - 4 | 319 | 59 | ||
| AK170757.1 | - | 3 - 4 | 365 | 59 | ||
| BC019524.1 | - | 3 - 4 | 427 | 4209097 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek6
Genomic Map