Assay Details
| Gene Symbol: Nek6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159631.1 | NP_001153103.1 | 7 - 8 | 754 | 83 | |
| NM_021606.3 | NP_067619.1 | 7 - 8 | 747 | 83 | ||
| GenBank mRNA | AF218847.1 | - | 7 - 8 | 749 | 83 | |
| AK004925.1 | - | 7 - 8 | 724 | 83 | ||
| AK011519.1 | - | 7 - 8 | 733 | 83 | ||
| AK029266.1 | - | 7 - 8 | 752 | 83 | ||
| AK075717.1 | - | 7 - 8 | 721 | 83 | ||
| AK086700.1 | - | 752 | 83 | |||
| AK089919.1 | - | 7 - 8 | 708 | 83 | ||
| AK170757.1 | - | 7 - 8 | 754 | 83 | ||
| BC019524.1 | - | 7 - 8 | 816 | 4209097 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek6
Genomic Map