Assay Details
| Gene Symbol: B4galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022305.4 | NP_071641.1 | 4 - 5 | 1695 | 78 | |
| GenBank mRNA | AK131721.1 | - | 4 - 5 | 798 | 78 | |
| AK132970.1 | - | 4 - 5 | 1696 | 78 | ||
| AK154393.1 | - | 4 - 5 | 1154 | 78 | ||
| AK167322.1 | - | 4 - 5 | 954 | 78 | ||
| BC053006.1 | - | 4 - 5 | 1156 | 5698356 | 78 | |
| D00314.1 | - | 4 - 5 | 1109 | 78 | ||
| J03880.1 | - | 4 - 5 | 1134 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B4galt1
Genomic Map