Assay Details
| Gene Symbol: Nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF284439.1 | - | 1 - 2 | 483 | 104 | |
| AK076788.1 | - | 119 | 104 | |||
| AK131631.1 | - | 1 - 2 | 369 | 104 | ||
| AK154815.1 | - | 1 - 2 | 98 | 104 | ||
| AK158616.1 | - | 1 - 2 | 389 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF284439.1, AK076788.1, AK131631.1, AK154815.1, AK158616.1 transcripts.
Set Membership: |
|

Nif3l1
Genomic Map