Assay Details
| Gene Symbol: Nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022988.2 | NP_075364.2 | 5 - 6 | 974 | 61 | |
| GenBank mRNA | AF284439.1 | - | 6 - 7 | 1451 | 61 | |
| AK003978.1 | - | 6 - 7 | 1082 | 61 | ||
| AK011670.1 | - | 6 - 7 | 991 | 61 | ||
| AK131631.1 | - | 6 - 7 | 1337 | 61 | ||
| AK154815.1 | - | 6 - 7 | 1066 | 61 | ||
| AK158616.1 | - | 6 - 7 | 1357 | 61 | ||
| BC065163.1 | - | 5 - 6 | 974 | 6404016 | 61 | |
| BC089475.1 | - | 6 - 7 | 901 | 30504614 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
neuron differentiation
positive regulation of transcription, DNA-templated
negative regulation of nucleic acid-templated transcription
nucleotide binding
GTP cyclohydrolase I activity
protein binding
GTP binding
transcription factor binding
hydrolase activity
identical protein binding
metal ion binding
neuron differentiation
positive regulation of transcription, DNA-templated
negative regulation of nucleic acid-templated transcription
nucleotide binding
GTP cyclohydrolase I activity
protein binding
GTP binding
transcription factor binding
hydrolase activity
identical protein binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100809 | AK076788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100809 | BC015257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100809 | U27457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100809 | AK146281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100809 | AK146859.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | BC065163.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | BC089475.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | AK131631.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | AK154815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | AK158616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | BC065163.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | BC089475.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | AK131631.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | AK154815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P479043 | AK158616.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417873 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419292 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313388 | AK080399.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313388 | XM_001472088.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313388 | XM_001477008.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158277 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27383 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | BC065163.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | BC089475.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | AK131631.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | AK154815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | AK158616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | BC065163.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | BC089475.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | AK131631.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | AK154815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P479043 | AK158616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100809 | AK076788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100809 | BC015257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100809 | U27457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100809 | AK146281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100809 | AK146859.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20258971 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00076842 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00076859 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04110359 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00076846 | - | - |
| Whole Mouse Genome G4122A | A_52_P181060 | BC065163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P181060 | AK131631.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P181060 | NM_022988.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | BC065163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | BC089475.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | AK131631.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | AK154815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | AK158616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | BC065163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | BC089475.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | AK131631.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | AK154815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479043 | AK158616.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110302 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00076842 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00855809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00855815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20258945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00452324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00640835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00378801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00855810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00378800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00855812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20258979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00077117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00377359 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10346538 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346540 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346541 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346539 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346537 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346542 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346543 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416931_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438728_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10346533 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10354832 | - | - |
| Mouse Genome 430 2.0 Array | 1416931_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438728_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438728_at | - | - |
| Mouse Genome 430 2.0 Array | 1416931_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65102_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65102_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100003511.mm.1 | NM_022988.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003511.mm.1 | TC0100000434.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100003510.mm.1 | NM_022988.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003510.mm.1 | TC0100000434.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416931_PM_at | - | - |
| Mouse Expression Set 430 | 1416931_at | - | - |
| Mouse Expression Set 430 | 1438728_PM_at | - | - |
| Mouse Expression Set 430 | 1438728_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416931_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416931_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416931_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416931_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438728_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438728_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 914692 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2760239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4560400 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2760239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2738837 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2717690 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2717690 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2738837 | - | - |
| mouse-6 v1.1 (Illumina) | scl17933.6.96_12-S | - | - |
| mouse-6 v1.1 (Illumina) | scl065102.6_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760239 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560400 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17933.6.96_12-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl065102.6_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560400 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2717690 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2738837 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2738837 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2717690 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl17933.6.96_12-S | - | - |

Nif3l1
Genomic Map