Assay Details
| Gene Symbol: Prnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126338.2 | NP_001119810.1 | 1 - 2 | 51 | 58 | |
| NM_001278257.1 | NP_001265186.1 | 1 - 2 | 51 | 58 | ||
| NM_023043.3 | NP_075530.1 | 1 - 2 | 51 | 58 | ||
| GenBank mRNA | AF192384.1 | - | 1 - 2 | 50 | 58 | |
| AF192385.1 | - | 1 - 2 | 50 | 58 | ||
| AK043023.1 | - | 1 - 2 | 52 | 58 | ||
| AK161043.1 | - | 1 - 2 | 52 | 58 | ||
| DQ832281.1 | - | 1 - 2 | 52 | 58 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF192384.1,AF192385.1,AK043023.1,AK161043.1,DQ832281.1,NM_001126338.2,NM_001278257.1,NM_023043.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081840 | BC025140.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081840 | DQ832281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081840 | AF192385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081840 | AK043023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081840 | AK161043.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186324 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580491 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156050 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129438 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081840 | BC025140.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081840 | DQ832281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081840 | AF192385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081840 | AK043023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081840 | AK161043.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177371 | BC025140.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177371 | DQ832281.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177371 | AF192385.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177371 | AK043023.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177371 | AF165165.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21637638 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129438 | - | - |
| Whole Mouse Genome G4122A | A_51_P177371 | BC025140.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177371 | DQ832281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177371 | AF192385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177371 | AK043023.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177371 | AF165165.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129438 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01606809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01606813 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01606835 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01606819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21637645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01695466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01991477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01738752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02104570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01695469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02258212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01372799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01606818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02258201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21637624 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10476322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476325 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425681_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10476321 | - | - |
| Mouse Genome 430 2.0 Array | 1425681_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425681_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26434_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26434_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200016129.mm.1 | NM_001126338.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016129.mm.1 | NM_001278257.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016129.mm.1 | NM_023043.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016129.mm.1 | TC0200002026.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200016133.mm.1 | NM_001278257.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016133.mm.1 | NM_001278258.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016133.mm.1 | NM_023043.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016133.mm.1 | TC0200002026.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425681_PM_a_at | - | - |
| Mouse Expression Set 430 | 1425681_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425681_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425681_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425681_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425681_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200440 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200440 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200440 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2648732 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2648732 | - | - |
| mouse-6 v1.1 (Illumina) | scl026434.3_264-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200440 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl026434.3_264-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200440 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2648732 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2648732 | - | - |

Prnd
Genomic Map