Assay Details
| Gene Symbol: Pkmyt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023058.3 | NP_075545.2 | 4 - 5 | 1395 | 77 | |
| GenBank mRNA | AF175892.1 | - | 4 - 5 | 1279 | 77 | |
| AK031787.1 | - | 3 - 4 | 2262 | 77 | ||
| AK158550.1 | - | 4 - 5 | 1396 | 77 | ||
| AK165164.1 | - | 4 - 5 | 1262 | 77 | ||
| BC025061.1 | - | 4 - 5 | 1118 | 2609421 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkmyt1
Genomic Map