Assay Details
| Gene Symbol: Tbata | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017407.1 | NP_001017407.1 | 1 - 2 | 145 | 71 | |
| NM_001017409.1 | NP_001017409.1 | 1 - 2 | 145 | 71 | ||
| NM_023064.3 | NP_075551.3 | 1 - 2 | 145 | 71 | ||
| GenBank mRNA | AF257502.1 | - | 1 - 2 | 145 | 71 | |
| AF257503.1 | - | 1 - 2 | 145 | 71 | ||
| AY243457.1 | - | 1 - 2 | 145 | 71 | ||
| EU622858.1 | - | 1 - 2 | 62 | 71 | ||
More Information
Set Membership: |
|

Tbata
Genomic Map