Assay Details
| Gene Symbol: Tbata | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017433.2 | NP_001017433.1 | 3 - 4 | 477 | 101 | |
| NM_023064.3 | NP_075551.3 | 2 - 3 | 395 | 101 | ||
| GenBank mRNA | AF257502.1 | - | 2 - 3 | 395 | 101 | |
| AF521592.1 | - | 2 - 3 | 408 | 101 | ||
| AJ249392.1 | - | 1 - 2 | 265 | 101 | ||
| AK088524.1 | - | 1 - 2 | 264 | 101 | ||
| AY243459.1 | - | 1 - 2 | 265 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbata
Genomic Map