Assay Details
| Gene Symbol: Ndnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023239.4 | NP_075728.1 | 1 | 266 | 113 | |
| GenBank mRNA | AF319979.1 | - | 1 | 144 | 113 | |
| AK010294.1 | - | 1 | 227 | 113 | ||
| AK017727.1 | - | 1 | 268 | 113 | ||
| AK018250.1 | - | 1 | 227 | 113 | ||
| AK049759.1 | - | 1 | 246 | 113 | ||
| AK076471.1 | - | 1 | 252 | 113 | ||
| AK131157.1 | - | 223 | 113 | |||
| BC034892.1 | - | 1 | 191 | 1494952 | 113 | |
| BC092289.1 | - | 1 | 228 | 4990134 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179905 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179903 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179902 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179906 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179907 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179904 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | BC034892.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | BC092289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | AK010294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | AK017727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | AK018250.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | BC034892.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | BC092289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | AK010294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | AK017727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333869 | AK018250.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153161 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146595 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | BC034892.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | BC092289.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | AK010294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | AK017727.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | AK018250.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | BC034892.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | BC092289.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | AK010294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | AK017727.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333869 | AK018250.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P196721 | AK131157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | BC034892.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | BC092289.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | AK010294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | AK017727.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | AK018250.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | BC034892.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | BC092289.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | AK010294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | AK017727.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P333869 | AK018250.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25179951 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153161 | - | - |
| Whole Mouse Genome G4122A | A_52_P196721 | AK131157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | BC034892.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | BC092289.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | AK010294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | AK017727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | AK018250.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | BC034892.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | BC092289.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | AK010294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | AK017727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333869 | AK018250.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111295 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153161 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06231146 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06554264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06554266 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179905 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179902 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179903 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179906 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179907 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25179904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25179946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06509909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06554266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06231138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06231143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06231131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25179950 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10564341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564339 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422554_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10564337 | - | - |
| Mouse Genome 430 2.0 Array | 1422554_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422554_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66647_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66647_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700029149.mm.1 | NM_023239.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029149.mm.1 | NM_023239.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029149.mm.1 | TC0700003415.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700029151.mm.1 | NM_023239.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029151.mm.1 | NM_023239.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029151.mm.1 | TC0700003415.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700029146.mm.1 | NM_023239.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029146.mm.1 | NM_023239.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029146.mm.1 | TC0700003415.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700029148.mm.1 | NM_023239.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029148.mm.1 | NM_023239.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700029148.mm.1 | TC0700003415.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422554_at | - | - |
| Mouse Expression Set 430 | 1422554_PM_at | - | - |
| Murine Genome U74v2 Set | 103220_PM_at | - | - |
| Murine Genome U74v2 Set | 103220_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422554_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422554_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422554_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422554_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 379041 | - | - |
| ABI Mouse Genome Survey Microarray | 932849 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6550458 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550458 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691441 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691441 | - | - |
| mouse-6 v1.1 (Illumina) | scl31245.1.43_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550458 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31245.1.43_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550458 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31245.1.43_5-S | - | - |

Ndnl2
Genomic Map