Assay Details
| Gene Symbol: Hspbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024172.3 | NP_077134.1 | 6 - 7 | 1050 | 63 | |
| GenBank mRNA | AF338351.1 | - | 5 - 6 | 988 | 63 | |
| AK013263.1 | - | 6 - 7 | 1041 | 63 | ||
| AK088469.1 | - | 6 - 7 | 1026 | 63 | ||
| AK160128.1 | - | 6 - 7 | 1046 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspbp1
Genomic Map