Assay Details
| Gene Symbol: Nrbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036293.2 | NP_001031370.1 | 1 - 2 | 185 | 87 | |
| GenBank mRNA | AK002810.1 | - | 1 - 2 | 187 | 87 | |
| AK004200.1 | - | 1 - 2 | 107 | 87 | ||
| AK189272.1 | - | 1 - 2 | 38 | 87 | ||
| BC019448.1 | - | 1 - 2 | 185 | 4212132 | 87 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK002810.1,BC019448.1,NM_001036293.2 transcripts.
Set Membership: |
|

Nrbf2
Genomic Map