Assay Details
| Gene Symbol: Aurkaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025338.4 | NP_079614.1 | 1 - 2 | 466 | 119 | |
| GenBank mRNA | AK002737.1 | - | 1 - 2 | 85 | 119 | |
| AK003442.1 | - | 1 - 2 | 90 | 119 | ||
| AK033452.1 | - | 1 - 2 | 73 | 119 | ||
| AK166049.1 | - | 1 - 2 | 70 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002737.1, AK003442.1, AK033452.1, AK166049.1, NM_025338.4 transcripts.
Set Membership: |
|

Aurkaip1
Genomic Map