Assay Details
| Gene Symbol: 1110001J03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025363.3 | NP_079639.1 | 1 - 2 | 148 | 73 | |
| GenBank mRNA | AK003231.2 | - | 1 - 2 | 169 | 73 | |
| AK008291.2 | - | 1 - 2 | 149 | 73 | ||
| AK009186.1 | - | 1 - 2 | 142 | 73 | ||
| BC059710.1 | - | 1 - 2 | 164 | 6539235 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24377801 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24377802 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24377799 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24377798 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406077 | BC059710.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406077 | AK008291.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406077 | AK009186.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406077 | NM_025363.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406077 | BC059710.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406077 | AK008291.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406077 | AK009186.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406077 | NM_025363.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513983 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99608 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117928 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406077 | BC059710.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406077 | AK008291.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406077 | AK009186.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406077 | NM_025363.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406077 | BC059710.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406077 | AK008291.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406077 | AK009186.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406077 | NM_025363.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24377778 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117928 | - | - |
| Whole Mouse Genome G4122A | A_51_P406077 | BC059710.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406077 | AK008291.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406077 | AK009186.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406077 | NM_025363.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406077 | BC059710.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406077 | AK008291.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406077 | AK009186.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406077 | NM_025363.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111939 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117928 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05168084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05168089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05168086 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05233475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24377802 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24377799 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24377801 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24377798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24377802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05772886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05012432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05462681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05405769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05168080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05168079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05427125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05168089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05012431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05168088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05661860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05781976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05872176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05661867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24377793 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10537396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537395 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416367_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10537394 | - | - |
| Mouse Genome 430 2.0 Array | 1416367_at | - | - |
| Mouse Genome 430 2.0 Array | 1416367_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66117_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66117_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600002885.mm.1 | TC0600000379.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600002885.mm.1 | NM_025363.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600002885.mm.1 | NM_025363.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600002886.mm.1 | TC0600000379.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600002886.mm.1 | NM_025363.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600002886.mm.1 | NM_025363.3 | Refseq |
| Mouse Expression Set 430 | 1416367_PM_at | - | - |
| Mouse Expression Set 430 | 1416367_at | - | - |
| Murine Genome U74v2 Set | 94907_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94908_r_at | - | - |
| Murine Genome U74v2 Set | 94908_PM_r_at | - | - |
| Murine Genome U74v2 Set | 94907_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416367_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416367_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416367_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416367_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 677462 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2570184 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2570184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002570184 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242707 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242707 | - | - |
| mouse-6 v1.1 (Illumina) | scl30187.2.1_182-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2570184 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30187.2.1_182-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2570184 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2958076 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2958076 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl30187.2.1_182-S | - | - |

1110001J03Rik
Genomic Map