Assay Details
| Gene Symbol: Tomm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025365.3 | NP_079641.1 | 2 - 3 | 586 | 119 | |
| GenBank mRNA | AK003291.1 | - | 2 - 3 | 248 | 119 | |
| AK010470.1 | - | 2 - 3 | 298 | 119 | ||
| AK019199.1 | - | 2 - 3 | 248 | 119 | ||
| BC037589.1 | - | 2 - 3 | 286 | 5007624 | 119 | |
| CA321392.1 | - | 2 - 3 | 170 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tomm6
Genomic Map