Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100040719.mm.1 |
NM_025402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040719.mm.1 |
NM_025402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040719.mm.1 |
TC1100004248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040723.mm.1 |
NM_025402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040723.mm.1 |
NM_025402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040723.mm.1 |
TC1100004248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040720.mm.1 |
NM_025402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040720.mm.1 |
NM_025402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040720.mm.1 |
TC1100004248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040718.mm.1 |
NM_025402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040718.mm.1 |
NM_025402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040718.mm.1 |
TC1100004248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040722.mm.1 |
NM_025402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040722.mm.1 |
NM_025402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040722.mm.1 |
TC1100004248.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040721.mm.1 |
NM_025402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040721.mm.1 |
NM_025402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040721.mm.1 |
TC1100004248.mm.1 |
MTA TC |