Assay Details
| Gene Symbol: Cd302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290660.1 | NP_001277589.1 | 2 - 3 | 532 | 117 | |
| NM_025422.4 | NP_079698.2 | 2 - 3 | 532 | 117 | ||
| GenBank mRNA | AI116405.1 | - | 2 - 3 | 532 | 117 | |
| AK002812.1 | - | 2 - 3 | 173 | 117 | ||
| AK004267.1 | - | 2 - 3 | 468 | 117 | ||
| BC005501.2 | - | 1 - 2 | 94 | 3489789 | 117 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC005501.2 transcripts.
Set Membership: |
|

Cd302
Genomic Map